Order Entry
Denmark
Orders LinkContactUsLinkComponent
Pharmaceutical exhausts

Pharmaceutical exhausts

Pharmacie fume hood, APA.145.090, 330 m³/h, 900×600×1450 mm, 890×440×930 mm
Katalog # 135-1773
Leverandør:   asecos
Pharmaceutical exhausts
Katalog # 135-1773
Leverandør:  asecos
Pharmaceutical exhausts
Pharmaceutical exhausts

Pharmacie fume hood, APA.145.090, 330 m³/h, 900×600×1450 mm, 890×440×930 mm

Price
20.870,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

For safe handling of chemicals, protection from hazardous vapours and potentially explosive atmospheres. Hazardous material workplace (pharmaceutical exhausts).

  • Easily mounted on a table top
  • Robust construction made of chemically resistant, antibacterial plastic laminated sheet steel
  • Exhaust air monitoring with visual and audible alarm

Standard equipment: exhaust air monitoring, 2 sockets, workplace lighting, acrylic glass sliding windows with continuously adjustable height, exhaust air fan. Accessories like media supplies, support frame and ventilation sets are available on request.

Standard equipment: exhaust air monitoring, 2 sockets, workplace lighting, acrylic glass sliding windows with coninuously adjustable height, exhaust air fan. Accessories like media supplies, support frame and ventilation sets are available on request.

Pressure drop with extraction air duct: 315 PA

 : Conform to DIN 12924 Part 4: 2012 CE. Made in Germany. Testing of the air equipment by recognised test institute

 : Delivery prices avaible upon request.

Specifikationer
  • Flow rate:
    330 m³/h
  • W×D×H ext.:
    900×600×1450 mm
  • Description:
    Pharmacie fume hood, APA.145.090
  • Seq:
    02
  • W×D×H int.:
    890×440×930 mm
  • Cat. No.:
    135-1773
  • Model:
    APA.145.090
  • Drain Ø:
    12 mm
  • Exhaust connection NW:
    100 mm
  • Weight:
    65 kg
  • Noise level:
    55 dB(A)