[EN]CAP SNAP-ON IN PE 13MM RUBBER/PTFE 1 * 100 ST
Catalog # BROW158125
Supplier: BROWN CHROMATOGRAPHY SUPPLIES
[EN]CAP SNAP-ON IN PE 13MM RUBBER/PTFE 1 * 100 ST
Catalog # BROW158125
Supplier: BROWN CHROMATOGRAPHY SUPPLIES
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
98.0-101.0% Ph. Eur.
Supplier: VWR Chemicals
[EN]SNAP CAP 8MM WITH RUBBER/PTFE SEPTA 1 * 100 ST
Supplier: BROWN CHROMATOGRAPHY SUPPLIES
Glass bottom microplates, SensoPlate™
Supplier: Greiner Bio-One