Order Entry
Denmark
ContactUsLinkComponent
lysate, cell/tissue lysate, raw 264.7 (mouse macrophage, abelson murine leukemia virus transformed) whole cell lysate (denatured), w hole cell lysate (denatured) 1 * 200 µG
Catalog # ABNOL024V1
Supplier:  Abnova
lysate, cell/tissue lysate, raw 264.7 (mouse macrophage, abelson murine leukemia virus transformed) whole cell lysate (denatured), w hole cell lysate (denatured) 1 * 200 µG
Catalog # ABNOL024V1
Supplier:  Abnova