Order Entry
Denmark
ContactUsLinkComponent
kit, epstein barr virus vca igm elisa kit, epstein barr virusvca igm elisa kit is intended for the detection of igm antibodies to eb v-vca in human serum or plasma. 1 * 1 KIT
Catalog # ABNOKA2255
Supplier:  Abnova
kit, epstein barr virus vca igm elisa kit, epstein barr virusvca igm elisa kit is intended for the detection of igm antibodies to eb v-vca in human serum or plasma. 1 * 1 KIT
Catalog # ABNOKA2255
Supplier:  Abnova