Order Entry
Denmark
ContactUsLinkComponent
OVERHEAD MIXER ROTAX 6.8, VARIABLE SPEED 0-30 RPM, DIGITAL DISPLAY, 6X2L OR 8X1L BOTTLES 1 * 1 items
Catalog # VELPF10600118
Supplier:  VELP SCIENTIFIC
undefined
OVERHEAD MIXER ROTAX 6.8, VARIABLE SPEED 0-30 RPM, DIGITAL DISPLAY, 6X2L OR 8X1L BOTTLES 1 * 1 items
Catalog # VELPF10600118
Supplier:  VELP SCIENTIFIC
Frequently Bought Together