Cover spectacles, uvex overspecs
Anit-UV (2-1,2), anti-scratch, anti-fog, anti-static, Blue/black, Clear PC
111-1651
: UVEX
The cover glasses (OTG) are suited for a range of applications and fit over most prescription spectacles. Whether as visitor spectacles or as overspecs for short-term tasks, they allow perfect vision due to the large panoramic lenses.
- Unrestricted peripheral vision
- Pressure-free fit through duo-flex side arms
- Side arm length adjustment
- Integrated brow guard
Weight: 39 g
EN 166
: EN 166
- Type:Anit-UV (2-1,2), anti-scratch, anti-fog, anti-static
- Colour:Blue/black
- Seq:02
- Lenses:Clear PC