VWR®, Cell Holders
Cell holder (reference), 1 position for 10 mm standard cells
Catalog # 634-0778
Supplier: VWR Collection
Jump to:
Product Details & Documents
Documents
Specifications
- Description:Cell holder (reference), 1 position for 10 mm standard cells
- Seq:04
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



