Order Entry
Denmark
Orders LinkContactUsLinkComponent
Inserts

Inserts

A 7/1 insert, perforated plate, for E 775/1, E 735/1 and E 701/1
7×7 mm perforations, 3 mm ridge, G 7825, G 7826, 543×473×1 mm
Catalog # 141-0492
Inserts
Catalog # 141-0492
Supplier:  
Inserts
Inserts

A 7/1 insert, perforated plate, for E 775/1, E 735/1 and E 701/1
7×7 mm perforations, 3 mm ridge, G 7825, G 7826, 543×473×1 mm

Discontinued
Product Details & Documents
Specifications
  • Description:
    A 7/1 insert, perforated plate, for E 775/1, E 735/1 and E 701/1
    7×7 mm perforations, 3 mm ridge
  • Seq:
    1
  • W×D×H:
    543×473×1 mm
  • For:
    G 7825, G 7826
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.