Cover spectacles, Squale
Welding, anti-scratch, Grey, Shade 5 PC
Catalog # BOLESQUWPCC5
Supplier: Bollé Safety
Jump to:
Product Details & Documents
Cover glasses (OTG) give excellent protection for wearers of prescription eyewear. Supplied with adjustable cord.
- Adjustable Nylon temples
- Moulded lens, brow guard and side shields
- Indirect ventilation
Weight: 43 g
Certifications: EN 166, EN 170
Specifications
- Type:Welding, anti-scratch
- Colour:Grey
- Lenses:Shade 5 PC