Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Diaphragm pump for drying train circulation, for 230 V units, 35 l/minute displacement capacity
Catalog # LBCP5248201
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP5248201
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Diaphragm pump for drying train circulation, for 230 V units, 35 l/minute displacement capacity
Price
16.660,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Beskrivelse:
    Diaphragm pump for drying train circulation, for 230 V units, 35 l/minute displacement capacity
  • Seq:
    27