Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Ampicillin

Glass bottle for solids, 25 g
Catalog # J60977.14
Health
Irritant
Ampicillin
Catalog # J60977.14
CAS Number:  69-53-4
Ampicillin
Ampicillin

Glass bottle for solids, 25 g

CAS Number:  69-53-4
Price
5.900,00 kr
Log in for availability
1 * 25 g
Product Details & Documents

CAS No.: 69-53-4 Molecular Formula: C16H19N3O4S Formula Weight: 349.40 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white Melting Point: 208° dec. Solubility: In 1 M NH4OH, 50 mg/ml Beilstein Reference: 1090925 Merck Reference: 14,586 MDL No.: MFCD00005175

  • CAS Number:69-53-4
  • Formula:C₁₆H₁₉N₃O₄S
  • MDL Number:MFCD00005175
  • Molecular Weight:349.4 g/mol
  • Merck Index:14,586
  • Melting Pt:208 °C (dec.)
  • EINECS:200-709-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications