Ampicillin
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 69-53-4 Molecular Formula: C16H19N3O4S Formula Weight: 349.40 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white Melting Point: 208° dec. Solubility: In 1 M NH4OH, 50 mg/ml Beilstein Reference: 1090925 Merck Reference: 14,586 MDL No.: MFCD00005175
- CAS Number:69-53-4
- Formula:C₁₆H₁₉N₃O₄S
- MDL Number:MFCD00005175
- Molecular Weight:349.4 g/mol
- Merck Index:14,586
- Melting Pt:208 °C (dec.)
- EINECS:200-709-7
Documents
Certificates
Search by Lot NumberALFAJ60977.06_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



