CHES, 0.5 M buffer solutions
CHES buffer solution, 0,5 M, 9.0, 250 mL
2-(Cyclohexylamino)ethanesulfonic acid, CHES.
CAS No.: 103-47-9
MDL No.: MFCD00003835
- Concentration:0,5 M
- Pk:250 mL
- Description:CHES buffer solution
- pH value:9.0
- Cat. No.:J63670.AK
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- MDL No.:MFCD00003835
- CAS No.:103-47-9



