Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)
TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer), Glass bottle, 2 L
Supplier: Thermo Fisher Scientific
Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)
Catalog # J63487.K3
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Physical Form: Liquid
MDL No.: MFCD00236358
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2 L
- Description:TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer)
- Cat. No.:J63487.K3
- Physical form:Liquid
- MDL No.:MFCD00236358
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



