Order Entry
Denmark
Orders LinkContactUsLinkComponent
Stacking buffer (4X)

Stacking buffer (4X)

Stacking Buffer, 4X, 500 mL
Catalog # J63450.AP
Stacking buffer (4X)
Catalog # J63450.AP
CAS Number:  
Stacking buffer (4X)
Stacking buffer (4X)

Stacking Buffer, 4X, 500 mL

CAS Number:  
Price
854,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents

Physical Form: Liquid

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Stacking Buffer, 4X
  • Cat. No.:
    J63450.AP
  • Physical form:
    Liquid

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.