Order Entry
Denmark
Orders LinkContactUsLinkComponent
Forskolin ≥98%

Forskolin ≥98%

Glass bottle for solids, 50 mg
Catalog # J63292.MF
Irritant
Forskolin ≥98%
Catalog # J63292.MF
CAS Number:  66575-29-9
Forskolin ≥98%
Forskolin ≥98%

Glass bottle for solids, 50 mg

CAS Number:  66575-29-9
Price
10.440,00 kr
Log in for availability
1 * 50 mg
Product Details & Documents

Purity: 98+% CAS No.: 66575-29-9 Molecular Formula: C22H34O7 Formula Weight: 410.51 Storage Temperature: -30°C to -10°C Physical Form: Solid Solubility: Soluble in DMSO or ethanol. Beilstein Reference: 1692716 Merck Reference: 14,2476 MDL No.: MFCD00082317

An adenylcyclase activator

Synonyms:  (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate
  • CAS Number:66575-29-9
  • Formula:C₂₂H₃₄O₇
  • MDL Number:MFCD00082317
  • Molecular Weight:410.51 g/mol
  • Merck Index:14,2476
  • EINECS:266-410-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg

Specifications