Order Entry
Denmark
Orders LinkContactUsLinkComponent
Bisacrylamide 2% in aqueous solution

Bisacrylamide 2% in aqueous solution

Glass bottle, 500 mL
Catalog # J63265.AP
Bisacrylamide 2% in aqueous solution
Catalog # J63265.AP
CAS Number:  110-26-9
Bisacrylamide 2% in aqueous solution
Bisacrylamide 2% in aqueous solution

Glass bottle, 500 mL

CAS Number:  110-26-9
Price
830,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS
  • CAS Number:110-26-9
  • Formula:(H₂C=CHCONH)₂CH₂
  • MDL Number:MFCD00008625
  • Molecular Weight:154.17 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:203-750-9
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications