HEPES, 0.5 M buffer solutions
HEPES buffer solution, 0,5 M, 8,5, Glass bottle, 250 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 7365-45-9
MDL No.: MFCD00006158
Specifications
Specifications
- Concentration:0,5 M
- Pack type:Glass bottle
- Pk:250 mL
- Description:HEPES buffer solution
- pH value:8,5
- Cat. No.:J63218.AK
- Physical form:Liquid
- MDL No.:MFCD00006158
- CAS No.:7365-45-9
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



