Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer), Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Catalog # J63931.K2
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00236357
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Description:TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)
- Cat. No.:J63931.K2
- Physical form:Liquid
- MDL No.:MFCD00236357
- Appearance:Colourless
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



