Buffers, potassium phosphate, 0,5 M solutions
Potassium phosphate, buffer solution, 0,5 M, 7,4, Glass bottle, 500 mL
Leverandør: Thermo Fisher Scientific
Buffers, potassium phosphate, 0,5 M solutions
Katalog # J61413.AP
Leverandør: Thermo Fisher Scientific
CAS nummer:
Gå til:
Product Details & Documents
CAS No.: 7778-77-0
Physical Form: Liquid
MDL No.: MFCD00011401
Documents
Specifikationer
Specifikationer
- Concentration:0,5 M
- Pack type:Glass bottle
- Pk:500 mL
- Description:Potassium phosphate, buffer solution
- pH value:7,4
- Cat. No.:J61413.AP
- Physical form:Liquid
- MDL No.:MFCD00011401
- CAS No.:7778-77-0
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



