Buffers, PIPES, 1,0 M solutions
PIPES buffer solution, 1,0 mol/L, 8,0, Glass bottle, 250 mL
Liquid
CAS No.: 5625-37-6
Appearance: Colorless
MDL No.: MFCD00006159
: Storage Temperature: 2 to 8 °C
- Concentration:1,0 mol/L
- Pack type:Glass bottle
- Pk:250 mL
- Description:PIPES buffer solution
- pH value:8,0
- Cat. No.:J60618.AK
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- MDL No.:MFCD00006159
- Appearance:Colourless
- CAS No.:5625-37-6



