Order Entry
Denmark
Orders LinkContactUsLinkComponent
TBE running buffer (10X)

TBE running buffer (10X)

TBE running buffer (10X), 2 L
Catalog # J62788.K3
TBE running buffer (10X)
Catalog # J62788.K3
CAS Number:  
TBE running buffer (10X)
TBE running buffer (10X)

TBE running buffer (10X), 2 L

CAS Number:  
Price
1.360,00 kr
Log in for availability
1 * 2 L
Product Details & Documents

Physical Form: Liquid
Appearance: Colourless

Specifications

Specifications

  • Pk:
    2 L
  • Description:
    TBE running buffer (10X)
  • Cat. No.:
    J62788.K3
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.