Buffers, potassium phosphate, 0,5 M solutions
Potassium phosphate, buffer solution, 0,5 M, 7,0, Glass bottle, 500 mL
Leverandør: Thermo Fisher Scientific
Buffers, potassium phosphate, 0,5 M solutions
Katalog # J60584.AP
Leverandør: Thermo Fisher Scientific
CAS nummer:
Gå til:
Product Details & Documents
CAS No.: 7778-77-0
Physical Form: Liquid
MDL No.: MFCD00011401
Documents
Specifikationer
Specifikationer
- Concentration:0,5 M
- Pack type:Glass bottle
- Pk:500 mL
- Description:Potassium phosphate, buffer solution
- pH value:7,0
- Cat. No.:J60584.AP
- Physical form:Liquid
- MDL No.:MFCD00011401
- CAS No.:7778-77-0



