Laemmli SDS sample buffer with pyronin Y, reducing (4X)
Laemmli SDS Sample Buffer, with PyroninY, 4X, 25 mL
Supplier: Thermo Fisher Scientific
Laemmli SDS sample buffer with pyronin Y, reducing (4X)
Catalog # J62115.AC
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Physical Form: Liquid
Appearance: Pink
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system
Documents
Specifications
Specifications
- Pk:25 mL
- Description:Laemmli SDS Sample Buffer, with PyroninY, 4X
- Cat. No.:J62115.AC
- Physical form:Liquid
- Appearance:Pink
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



