Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

HEPES, 0.5 M buffer solutions

HEPES buffer solution, 0,5 M, 7,5, Glass bottle, 100 mL
  J61275.AE
HEPES, 0.5 M buffer solutions
  J61275.AE
 :  
HEPES, 0.5 M buffer solutions
HEPES, 0.5 M buffer solutions

HEPES buffer solution, 0,5 M, 7,5, Glass bottle, 100 mL

 :  
 
930,00 kr
   
1 * 100 mL
 

 

  • Concentration:
    0,5 M
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    HEPES buffer solution
  • pH value:
    7,5
  • Cat. No.:
    J61275.AE
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00006158
  • CAS No.:
    7365-45-9