Order Entry
Denmark
Orders LinkContactUsLinkComponent
Stripping buffer-2 (4X)

Stripping buffer-2 (4X)

Stripping buffer, 4X, 500 mL
Catalog # J62023.AP
Stripping buffer-2 (4X)
Catalog # J62023.AP
CAS Number:  
Stripping buffer-2 (4X)
Stripping buffer-2 (4X)

Stripping buffer, 4X, 500 mL

CAS Number:  
Price
958,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Stripping buffer, 4X
  • Cat. No.:
    J62023.AP
  • Physical form:
    Liquid

Specifications