Buffer, ACES, 0,5 M solution
ACES, 0,5M buffer solution, pH 6,5, Glass bottle, 100 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 7365-82-4
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00008030
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Description:ACES, 0,5M buffer solution, pH 6,5
- Cat. No.:J60366.AE
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- MDL No.:MFCD00008030
- Appearance:Colourless
- CAS No.:7365-82-4
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



