Buffers, potassium phosphate, 0,5 M solutions
Potassium phosphate, buffer solution, 0,5 M, 7,2, Glass bottle, 250 mL
CAS No.: 7778-77-0
Physical Form: Liquid
MDL No.: MFCD00011401
- Concentration:0,5 M
- Pack type:Glass bottle
- Pk:250 mL
- Description:Potassium phosphate, buffer solution
- pH value:7,2
- Cat. No.:J60326.AK
- Physical form:Liquid
- MDL No.:MFCD00011401
- CAS No.:7778-77-0



