ADA, 0,2 M Buffer Solutions
ADA buffer solution, 0,2 M, 7,5, Glass bottle, 100 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
N-(2-Acetimido)iminodiacetic acid monosodium salt (ADA) buffer solutions PH values 6,0 to 7,5.
CAS No.: 7415-22-7
Appearance: Colourless
MDL No.: MFCD00065483
Caution: Storage Temperature: 2 to 8°C
Documents
Specifications
Specifications
- Concentration:0,2 M
- Pack type:Glass bottle
- Pk:100 mL
- Description:ADA buffer solution
- pH value:7,5
- Cat. No.:J62424.AE
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- MDL No.:MFCD00065483
- Appearance:Colourless
- CAS No.:7415-22-7



