Order Entry
Denmark
Orders LinkContactUsLinkComponent
TE buffer, pH 7,4, RNAse free

TE buffer, pH 7,4, RNAse free

TE buffer, RNAse free, 7,4, 250 mL
Catalog # J60234.AK
TE buffer, pH 7,4, RNAse free
Catalog # J60234.AK
CAS Number:  
TE buffer, pH 7,4, RNAse free
TE buffer, pH 7,4, RNAse free

TE buffer, RNAse free, 7,4, 250 mL

CAS Number:  
Price
1.360,00 kr
Log in for availability
1 * 250 mL
Product Details & Documents

TE buffer is a commonly used buffer solution in procedures involving DNA or RNA. It is used for storing and diluting DNA and RNA. Treatment with TE buffer enhances the staining intensity of antibodies in the immunohistochemical detection of certain proteins.

Physical Form: Liquid
Appearance: Clear
MDL No.: MFCD00236359

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    TE buffer, RNAse free
  • pH value:
    7,4
  • Cat. No.:
    J60234.AK
  • Storage temperature:
    RT
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00236359
  • Appearance:
    Clear

Specifications