HEPES, 0.5 M buffer solutions
HEPES buffer solution, 0,5 M, 8,0, Glass bottle, 250 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 7365-45-9
MDL No.: MFCD00006158
Documents
Specifications
Specifications
- Concentration:0,5 M
- Pack type:Glass bottle
- Pk:250 mL
- Description:HEPES buffer solution
- pH value:8,0
- Cat. No.:J63002.AK
- Physical form:Liquid
- MDL No.:MFCD00006158
- CAS No.:7365-45-9



