Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Phosphate, 0,5M buffer solution, pH 8,0

Phosphate buffer soln, 0,5 M, 8,0, 250 mL
  J61722.AK
Phosphate, 0,5M buffer solution, pH 8,0
  J61722.AK
 :  
Phosphate, 0,5M buffer solution, pH 8,0
Phosphate, 0,5M buffer solution, pH 8,0

Phosphate buffer soln, 0,5 M, 8,0, 250 mL

 :  
 
1.080,00 kr
   
1 * 250 mL
 

CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527

 

 

  • Koncentration:
    0,5 M
  • Pakn:
    250 mL
  • Beskrivelse:
    Phosphate buffer soln
  • pH-værdi:
    8,0
  • Cat. No.:
    J61722.AK
  • CAS-nr:
    7558-80-7
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00003527