Buffer, BIS-TRIS propane, 0,2 M solution, pH 6,5
BIS-TRIS propane buffer solution pH 6,5 (0,5 mol/l), Glass bottle, 250 mL
CAS No.: 64431-96-5
Physical form: liquid
MDL No.: MFCD00004689
- Pack type:Glass bottle
- Pk:250 mL
- Description:BIS-TRIS propane buffer solution pH 6,5 (0,5 mol/l)
- Cat. No.:J62354.AK
- Physical form:Liquid
- MDL No.:MFCD00004689
- CAS No.:64431-96-5



