Phosphate, 0,5M buffer solution, pH 9,0
Phosphate buffer soln, 0,5 M, 9,0, 250 mL
Supplier: Thermo Fisher Scientific
Phosphate, 0,5M buffer solution, pH 9,0
Catalog # J62345.AK
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527
Documents
Specifications
Specifications
- Concentration:0,5 M
- Pk:250 mL
- Description:Phosphate buffer soln
- pH value:9,0
- Cat. No.:J62345.AK
- Physical form:Liquid
- MDL No.:MFCD00003527
- CAS No.:7558-80-7



