MOPSO, 0,2 M buffer soln., pH 7,0
MOPSO buffer solution, 0,2 M, 7,0, 100 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 79803-73-9
Physical Form: Liquid
MDL No.: MFCD00070007
Documents
Specifications
Specifications
- Concentration:0,2 M
- Pk:100 mL
- Description:MOPSO buffer solution
- pH value:7,0
- Cat. No.:J61469.AE
- Physical form:Liquid
- MDL No.:MFCD00070007
- CAS No.:79803-73-9



