Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Accessories for Loopster basic and digital

Tube attachment for 48×1.5 ml tubes with Ø10 mm
  444-0850
 :   IKA
Accessories for Loopster basic and digital
  444-0850
 :  IKA
Accessories for Loopster basic and digital
Accessories for Loopster basic and digital

Tube attachment for 48×1.5 ml tubes with Ø10 mm

 
2.060,00 kr
   
1 * 1 Items
 
 
  • Description:
    Tube attachment for 48×1.5 ml tubes with Ø10 mm
  • Seq:
    03
Accessories

 

  •  
    Description
     
     
  • 444-0952
    Spare clips with Ø25 mm for use with Universal disc
       
    1 * 12 Items - 384,00 kr

 

  •  
    Description
     
     
  • 444-0857
    Spare clips with Ø12 mm for use with Universal disc
       
    1 * 12 Items - 384,00 kr

 

  •  
    Description
     
     
  • 444-0856
    Spare clips with Ø10 mm for use with Universal disc
       
    1 * 12 Items - 384,00 kr

 

  •  
    Description
     
     
  • 444-0953
    Spare clips with Ø28 mm for use with Universal disc
       
    1 * 12 Items - 384,00 kr

 

  •  
    Description
     
     
  • 444-0951
    Spare clips with Ø16 mm for use with Universal disc
       
    1 * 12 Items - 384,00 kr

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.