Accessories for Loopster basic and digital
Tube attachment for 24×5 ml tubes with Ø12 mm
Catalog # 444-0851
Supplier: IKA
Jump to:
Product Details & Documents
Specifications
- Description:Tube attachment for 24×5 ml tubes with Ø12 mm
- Seq:04
Accessories
Product Information
- Catalog #AvailabilityPrice
- Spare clips with Ø25 mm for use with Universal discSpare clips with Ø25 mm for use with Universal disc444-0952Log in for availability1 * 12 Items - 384,00 kr
Product Information
- Catalog #AvailabilityPrice
- Spare clips with Ø12 mm for use with Universal discSpare clips with Ø12 mm for use with Universal disc444-0857Log in for availability1 * 12 Items - 384,00 kr
Product Information
- Catalog #AvailabilityPrice
- Spare clips with Ø10 mm for use with Universal discSpare clips with Ø10 mm for use with Universal disc444-0856Log in for availability1 * 12 Items - 384,00 kr
Product Information
- Catalog #AvailabilityPrice
- Spare clips with Ø28 mm for use with Universal discSpare clips with Ø28 mm for use with Universal disc444-0953Log in for availability1 * 12 Items - 384,00 kr
Product Information
- Catalog #AvailabilityPrice
- Spare clips with Ø16 mm for use with Universal discSpare clips with Ø16 mm for use with Universal disc444-0951Log in for availability1 * 12 Items - 384,00 kr
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



