PROCARE UNIVERSAL 61 3 x 2KG 1 * 3 items
Catalog # MIEL09195780
Supplier: MIELE
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Shoe covers, Tyvek® 500, models POS0 / POSA
Supplier: DuPont
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
DETERGENT EDISONITE UNIV POWDER CPD 5KG 1 * 1 items
Supplier: Brand
Hydrochloric acid 0.1 mol/l (0.1 N), AVS TITRINORM Reag. Ph. Eur. volumetric solution
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection