Order Entry
Denmark
ContactUsLinkComponent
ELECTROPHORESIS COMB f. 28 SAMPLES MC Thickness:,,2 mm Sample volume:,,23 µl 1 * 1 items
Catalog # CONSEHS1300CMT2820
Supplier:  CONSORT
ELECTROPHORESIS COMB f. 28 SAMPLES MC Thickness:,,2 mm Sample volume:,,23 µl 1 * 1 items
Catalog # CONSEHS1300CMT2820
Supplier:  CONSORT
Frequently Bought Together