affinity chromatography of glucose binding proteins 1 * 1 items
Catalog # EDVO277
Supplier: EDVOTEK
Frequently Bought Together
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Isooctane (Reag. Ph. Eur.) for analysis, ACS 1 * 2,5 L
Supplier: PanReac AppliChem
Bromoform 96% stabilised
Supplier: Thermo Fisher Scientific
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥98.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Lithium perchlorate trihydrate 99%, extra pure
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
Copper(II) sulphate pentahydrate
Supplier: Hach
What's in my lunch? quantitative milk allergy ELISA kit
Supplier: EDVOTEK
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
Tetrachloroethylene ≥99%
Supplier: Thermo Fisher Scientific