Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sample containers, with screw cap, universal, WHEATON®
Sample containers, with screw cap, universal, WHEATON®
30 ml, Universal container, plain label, sterile, Translucent, PP/PE
Catalog # 216-0983
Supplier:   DWK Life Sciences
Sample containers, with screw cap, universal, WHEATON®
Catalog # 216-0983
Supplier:  DWK Life Sciences
Sample containers, with screw cap, universal, WHEATON®
Sample containers, with screw cap, universal, WHEATON®
30 ml, Universal container, plain label, sterile, Translucent, PP/PE
Price
1.080,00 kr
Log in for availability
1 * 400 Items
Product Details & Documents
PP translucent or PS clear, with PE screw cap.
  • Manufactured from FDA approved virgin polystyrene or polypropylene
  • PS offers good thermal and chemical resistance
  • Supplied sterile - for in vitro use only
  • All materials are non cytotoxic
Straight walled containers ideal for sample collection, storage and transport. Supplied with leakproof, flow seal closure. Available pre-filled with boric acid for urine samples.
Certifications: CE marked in accordance with European Directive 98/79/EC
All containers are leak tested to British Standard BS5213:1975(1989)
Packaging: Packed in inner bags of 50.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • Type:
    Universal container, plain label, sterile
  • Capacity:
    30 ml
  • Colour:
    Translucent
  • Material:
    PP/PE