Order Entry
Denmark
ContactUsLinkComponent
Additional pt 100, positioned flexibly in chamber or load, for temperature measurement, temperature can, if required, be indicated o n the display, recorded in the ring store for ufplus 1 * 1 items
Catalog # MEMMUF30PLUSH8
Supplier:  MEMMERT
Additional pt 100, positioned flexibly in chamber or load, for temperature measurement, temperature can, if required, be indicated o n the display, recorded in the ring store for ufplus 1 * 1 items
Catalog # MEMMUF30PLUSH8
Supplier:  MEMMERT
Frequently Bought Together