ClO2- Chlorites 1000 mg/l in H2O for IC 1 * 250 mL
Catalog # ACSDH028.01O.L25
Supplier: CPACHEM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Crucibles, Porcelain
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Sulphuric acid 95,0 - 97,0% 95.0-97.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals