Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Electrode Cables
Electrode Cables
Electrode cable for plug in head U/plug F, 2×2 mm B, for connecting electrodes with Metrohm plug-in head U to Metrohm instruments (socket F), 1 m
Catalog # METR6.2104.600
Supplier:   METROHM
Electrode Cables
Catalog # METR6.2104.600
Supplier:  METROHM
Electrode Cables
Electrode cable for plug in head U/plug F, 2×2 mm B, for connecting electrodes with Metrohm plug-in head U to Metrohm instruments (socket F), 1 m
Price
5.170,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Beskrivelse:
    Electrode cable for plug in head U/plug F, 2×2 mm B, for connecting electrodes with Metrohm plug-in head U to Metrohm instruments (socket F)
  • Længde:
    1 m
  • Seq:
    12