Order Entry
Denmark
ContactUsLinkComponent
Reusable freezing core for use in CoolBox XT and CoolBox 2XT systems freeze in -20 °C freezer, use for up to 12 hours of benchtop freezing 1 * 1 items
Catalog # 216-0954
Supplier:  BIOCISION
undefined
Reusable freezing core for use in CoolBox XT and CoolBox 2XT systems freeze in -20 °C freezer, use for up to 12 hours of benchtop freezing 1 * 1 items
Catalog # 216-0954
Supplier:  BIOCISION
Frequently Bought Together