Order Entry
Denmark
Orders LinkContactUsLinkComponent
Volumetric flasks, class B, PP

Volumetric flasks, class B, PP

500 ml, GL25, 270 mm, 0,50 ± ml
Catalog # VITL675891
Supplier:   VITLAB
Volumetric flasks, class B, PP
Catalog # VITL675891
Supplier:  VITLAB
Volumetric flasks, class B, PP
Volumetric flasks, class B, PP

500 ml, GL25, 270 mm, 0,50 ± ml

Price
328,00 kr
Log in for availability
1 * 2 Items
Product Details & Documents

PP, highly transparent, with PP screw cap.

  • With ring mark individually calibrated to ‘In’
  • Thermal stress up to 60 °C does not permanently exceed the tolerance limits
  • To preserve markings, cleaning at no higher than 60 °C is recommended

Class B tolerances according to DIN EN ISO 1042

Specifications
  • Thread:
    GL25
  • Capacity:
    500 ml
  • Height:
    270 mm
  • Tolerance:
    0,50 ± ml