Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tween® 20 (Polysorbate)

Tween® 20 (Polysorbate)

Glass bottle, 25 g
Catalog # TCIAT0543-25G
Supplier:   TCI
Tween® 20 (Polysorbate)
Catalog # TCIAT0543-25G
Supplier:  TCI
CAS Number:  9005-64-5
Tween® 20 (Polysorbate)

Glass bottle, 25 g

CAS Number:  9005-64-5
Price
140,00 kr
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Polyoxyethylene sorbitan monolaurate
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Molecular Weight:1227.53 g/mol
  • Density:1,1 g/cm³ (20 °C)
  • Flash Pt:297°C
  • Storage Temperature:Ambient
  • EINECS:500-018-3
Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow clear liquid
  • Saponification Value:
    43.0 to 53.0
  • Acid Value:
    max. 3.0

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.