Order Entry
Denmark
Orders LinkContactUsLinkComponent
Thymolphthalein

Thymolphthalein

Glass bottle, 25 g
Catalog # TCIAT0237-25G
Supplier:   TCI
Thymolphthalein
Catalog # TCIAT0237-25G
Supplier:  TCI
CAS Number:  125-20-2
Thymolphthalein
Thymolphthalein

Glass bottle, 25 g

CAS Number:  125-20-2
Price
378,00 kr
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
  • CAS Number:125-20-2
  • Formula:C₂₈H₃₀O₄
  • MDL Number:MFCD00005909
  • Molecular Weight:430.54 g/mol
  • Merck Index:14,09401
  • Melting Pt:253 °C
  • Storage Temperature:Ambient
  • EINECS:204-729-7
Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Absorbance:
    min. 0.350 (5mg/L, pH10.5, 592.0 to 596.0 nm)
  • Solubility in EtOH (50vol%):
    transparency
  • Drying loss:
    max. 1.0 %
  • Transition interval (pH):
    (colorless)8.6 to 10.5 (blue)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.