Thymolphthalein
Glass bottle, 1 g
TCIAT0237-1G
: TCI
: 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
- :125-20-2
- :C₂₈H₃₀O₄
- :MFCD00005909
- :430.54 g/mol
- :14,09401
- :253 °C
- :Ambient
- :204-729-7
- Appearance:White to Orange to Green powder to crystal
- Absorbance:min. 0.350 (5mg/L, pH10.5, 592.0 to 596.0 nm)
- Solubility in EtOH (50vol%):transparency
- Drying loss:max. 1.0 %
- Transition interval (pH):(colorless)8.6 to 10.5 (blue)
- Pack type:Glass bottle
- Pk:1 g



