Span® 80
Glass bottle, 500 g
Catalog # TCIAS0060-500G
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: Arlacel 80, Sorbitan monooleate
- CAS Number:1338-43-8
- Formula:C₂₄H₄₄O₆
- MDL Number:MFCD00080948
- Molecular Weight:428.61 g/mol
- Density:1 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-665-4
Specifications
Specification Test Results
- Appearance:Yellow to Amber to Dark purple clear liquid
- Saponification Value:142.0 to 189.0
- Acid Value:max. 10.0
Specifications
- Pack type:Glass bottle
- Pk:500 g
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



